What should I do if the Recovery Solution does not come out from Ion OneTouch DL?
The Ion OneTouch DL cannot be repaired. However, you may order an Ion OneTouch DL Upgrade Kit and exchange some parts which may fix the problem.