4469949 - FAQs
4469949 - FAQs
View additional product information for 4469949 - FAQs (4469949)
2 product FAQs found
What is the picture on the touchscreen during an Ion Proton run that looks like a bubble moving?
The touchscreen shows the voltage measured on the chip and there will be some contrast according the signal intensity which may look like bubbles.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Can I use the same oil between the Ion Proton OT2 200 Kit and the Ion Proton OT2 200 Kit v2?
No, you cannot. Please exchange the oil when switching between different kits. Please contact Technical Support for detailed information when switching kits.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.