tert-Butanol, 99.5%, for analysis
tert-Butanol, 99.5%, for analysis
tert-Butanol, 99.5%, for analysis
Thermo Scientific Chemicals

tert-Butanol, 99.5%, for analysis

CAS: 75-65-0 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number 447010010
Price (USD)
113.00
Each
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
113.00
Each
Chemical Identifiers
CAS75-65-0
IUPAC Name2-methylpropan-2-ol
Molecular FormulaC4H10O
InChI KeyDKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILESCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Boron (B)=<0.02 ppm
Magnesium (Mg)=<0.1 ppm
Nickel (Ni)=<0.02 ppm
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only