POROS™ GoPure™ XQ Pre-packed Column, 1.2 x 20 cm, 22.6 mL

Catalog number: 4470477

 Related applications: Chromatography

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

POROS™ GoPure™ pre-packed columns combine the flexibility of a traditional packed column with the convenience and reproducibility of a pre-packed ready-to-use column solution. GoPure™ pre-packed columns speed facility turnaround time by eliminating column qualification testing and the need for equipment cleaning and change-out. In addition, the columns are comparable in cost to packing a traditional column. GoPure™ pre-packed columns are packed and qualified in a dedicated clean room and are supported by numerous validation studies.

The POROS™ XQ Strong Anion Exchange resin in this column provides for high-capacity and high-resolution anion exchange chromatography with high salt tolerance.

Multi-product facilities requiring flexible, efficient, and cost-effective operations will find GoPure™ pre-packed columns fill a gap in downstream purification unit operations.

GoPure™ pre-packed columns are:
• Convenient—ready-to-run columns speed process setup and facility turnaround time
• Flexible design—bed heights available across multiple diameters, allowing columns to be tailored to process requirements
• Simplified scalability—qualified, reproducible pre-packed column simplifies process scale up or process modeling
• Qualified for use—columns packed in a dedicated clean room and include a column test certificate

Convenience and Performance Delivered
GoPure™ pre-packed columns are available in 1.2, 8.0, and 20 cm diameters, each with multiple bed heights that equate to a column volume range of 5 mL to 6.3 L. The simple design is reliable and sanitary. Moreover, the column construction materials meet regulatory requirements for toxicity and being animal origin-free. The columns arrive pre-qualified, sanitized, and stored in a bacteriostatic solution. They are shipped with a column test certificate that includes column efficiency, asymmetry, and permeability measurements.

GoPure™ pre-packed columns significantly reduce the time required to prepare for chromatography unit operations. In addition, GoPure™ columns can be campaign-dedicated, eliminating the time usually required to perform and document a cleaning validation. Depending on the facility workload, this time-saving feature can help to expand capacity without capital investments.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Binding Strength: 5% breakthrough capacity > 140 mg/mL BSA in the presence of up to 6.5 mS/cm NaCl,
Column Type: Ion Exchange
Format: Prepacked Column
Immobilized Phase: Strong anion exchange
Product Line: POROS®
Product Size: 1 column

Contents & storage

Store at room temperature.

Documents

Manuals & protocols