n-Butyl acetate, 99.5%, for biochemistry, AcroSeal™
n-Butyl acetate, 99.5%, for biochemistry, AcroSeal™
n-Butyl acetate, 99.5%, for biochemistry, AcroSeal™
n-Butyl acetate, 99.5%, for biochemistry, AcroSeal™
Thermo Scientific Chemicals

n-Butyl acetate, 99.5%, for biochemistry, AcroSeal™

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number 447050010
Price (EUR)
85,90
Each
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (EUR)
85,90
Each
Add to cart
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.01 %
Color scale=<15 APHA
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
RUO – Research Use Only