Product Image
Applied Biosystems™
GeneMapper™ ID-X Software Hotfix_20110620
Kit includes a Hotfix_20110620 CD for GeneMapper™ ID-X Software v1.0.1, 1.1 and 1.2 to fix installation and login problems thatRead more
Have Questions?
Catalog NumberQuantity
44706851 CD
Catalog number 4470685
Price (USD)
0.00
Each
Quantity:
1 CD
Price (USD)
0.00
Each
Kit includes a Hotfix_20110620 CD for GeneMapper™ ID-X Software v1.0.1, 1.1 and 1.2 to fix installation and login problems that occur on systems with certain Media Access Control (MAC) addresses. Release Note documentation included.
Multiple installations can be updated with a single Hotfix_20110620 CD.
This Kit includes GeneMapper™ ID-X v1.2 Client Upgrade CD for GeneMapper™ ID-X v1.0, 1.0.1, v1.1. and v1.1.1 Client installation upgrade. Documentation is not included.
GeneMapper™ ID-X Software v1.2 is a powerful data analysis software package designed to significantly reduce the amount of time required to perform sizing, allele calling, mixture analysis and statistical calculations on all types of forensic samples.
The new GeneMapper™ ID-X Software v1.2 was built upon the functionality of the GeneMapper™ ID-X Software v1.1 with the capability to support Applied Biosystems™ 3500 Series Genetic Analyzer for HID. This new version software has the expanded new features including new HID 3500 file format support, Vista operating system support, normalization, and 6-dye capability.
• Powerful, easy to use data analysis tool designed to increase lab productivity
• Comprehensive Expert System capability for analysis of forensic databasing samples delivered by automated data assessment functionality and efficient manual review tools
• Complete Expert Assistant solution for analysis of forensic casework samples combining an efficient suite of manual review features with an integrated mixture analysis tool
• Quality control functionality allows rapid allele match comparisons and concordance searches within a data set.
• Extensive security, auditing, and e-signature capabilities help protect data integrity and control technical records.
• Multi-user database configuration stores your projects in a central location and facilitates information exchange.
• Command Line Interface – the ability to access GeneMapper™ ID-X v1.1 Software automatically (without human intervention) streamlines its integration with LIMS installations and other applications in an existing laboratory infrastructure.
Automated Data Assessment
GeneMapper™ ID-X Software v1.2 is designed specifically to fulfill the requirements of both Expert System and Expert Assistant software. A combination of sophisticated automated data assessment processes and efficient manual review tools deliver comprehensive expert system capability. This provides optimal interaction between the analyst and the data and significantly reducing the amount of analysis time required for single source samples. New features such as the Analysis Requirements check, Allelic Ladder Quality Assessment, Improved Quality Value System and the Analysis Summary analyze samples and segregate them into pass or review categories based on your thresholds. Once segregated, you can easily conduct any necessary manual review of samples.
Despite its capabilities to automate and streamline the analysis of single source samples, an Expert System is as yet unable to make the final analysis decision for most forensic casework samples and in particular for those containing mixtures. GeneMapper™ ID-X Software v1.2 harness the automated, rule-based, subject-specific knowledge to act as an Expert Assistant to the forensic analyst, simplifying much of the analysis process and empowering the analyst to make key interpretation decisions. A mixture analysis tool determines the number of contributors to a mixture and efficiently manages the resulting possible genotypes. This enables the forensic analyst to interpret mixtures and generate accurate statistical calculations.
Manual Tools Improve Data Review
Data analysis efficiencies offered by the automated data review processes have been further enhanced through the introduction of a new suite of manual review tools. These developments focus around the plot window providing quick and easy access to all aspects of the data, thus simplifying the data review process. Colored marker header bars indicate which markers within a sample have failed to meet the user defined thresholds and the new Process Quality Value (PQV) Details window details which flags have been fired and how much the observed value differs from the threshold. In addition, expanded labeling options are available to distinguish alleles from artifacts and reviewers can directly access raw data for further troubleshooting. Electronic/peer review of the data is enhanced with the review and manual acceptance of marker and sample level PQVs along with a detailed label edit table display.
For further troubleshooting, reviewers can also directly access raw data. The ability to review and manually accept markers and sample-level PQVs in a detailed label edit table display enhances electronic/peer data review.
A Complete, Verified System
GeneMapper™ ID-X Software is an integral component of our complete system of instruments, reagents, software, and services for human-identity applications. It has undergone extensive verification testing to ensure robust design features, performance, and reproducibility. When you partner with us, you gain access to the most extensive Human Identification resource network in the industry. Our dedicated team of industry-leading experts – with extensive forensic product development experience – worked closely with the forensic community to develop GeneMapper™ ID-X Software. The software is part of a system that is supported by on-site technical assistance from skilled Field Applications Support Scientists and Field Service Engineers.
Monitor
Recommended Configuration (Full Installation):
• 1024 x 768 pixel resolution.
• 19-inch color monitor (enables greater viewing of data).
For Research Use Only. Not for use in diagnostics procedures.
Specifications
For Use With (Equipment)3500 Genetic Analyzer
Operating SystemWindows 7, Windows XP
Product TypeGeneMapper ID-X Software Hotfix
Quantity1 CD
Shipping ConditionRoom Temperature
Software CategoryHotfix
Product LineGeneMapper
Software TypeGeneMapper
Unit SizeEach
Frequently asked questions (FAQs)
Can you help me troubleshoot Example #4: Export Sample Plot to PDF with the -splitfile argument in the GeneMapperID-X Software Administrators Guide, Appendix A: Operating the Software from a Command Line Interface, page 108?
Unfortunately, there is no training or support for the GeneMapperID-X command line features.
We recommend referencing the GeneMapper 5 Installation and Administration User Guide, Appendix B for supported command line features and working with your informatics team for additional support.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.