TaqMan™ OpenArray™ Genotyping Training Plate, QuantStudio™ 12K Flex plate - FAQs
TaqMan™ OpenArray™ Genotyping Training Plate, QuantStudio™ 12K Flex plate - FAQs
View additional product information for TaqMan™ OpenArray™ Genotyping Training Plate, QuantStudio™ 12K Flex plate - FAQs (4471225)
1 product FAQs found
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?
Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.