Propionic acid, ACS reagent
Propionic acid, ACS reagent
Propionic acid, ACS reagent
Thermo Scientific Chemicals

Propionic acid, ACS reagent

CAS: 79-09-4 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number 447235000
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Chemical Identifiers
CAS79-09-4
IUPAC Namepropanoic acid
Molecular FormulaC3H6O2
InChI KeyXBDQKXXYIPTUBI-UHFFFAOYSA-N
SMILESCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.15 %
Color scale=<20 APHA
Titration with NaOH>=99.5 %
Residue after evaporation=<0.01 %
Oxidisable substance (HCOOH)=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only