Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Potassium phosphate, dibasic, 99%, for HPLC, anhydrous
Thermo Scientific Chemicals

Potassium phosphate, dibasic, 99%, for HPLC, anhydrous

CAS: 7758-11-4 | HK2O4P | 174.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
447362500250 gPlastic bottle
Catalog number 447362500
Price (USD)
254.65
Online exclusive
283.00
Save 28.35
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
254.65
Online exclusive
283.00
Save 28.35
Each
Chemical Identifiers
CAS7758-11-4
IUPAC Namedipotassium hydrogen phosphate
Molecular FormulaHK2O4P
InChI KeyZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES[K+].[K+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or granules
Loss on drying=<1 %
Titration with HClO4>=98.5 %
pH8.5 to 9.6 (10 % in water)
UVat 230 nm A: =<0.15
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only