Ammonium dihydrogen phosphate, 99%, for HPLC
Ammonium dihydrogen phosphate, 99%, for HPLC
Ammonium dihydrogen phosphate, 99%, for HPLC
Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, 99%, for HPLC

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
447382500250 gPlastic bottle
Catalog number 447382500
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Chemical Identifiers
CAS7722-76-1
IUPAC Namephosphoric acid amine
Molecular FormulaH6NO4P
InChI KeyLFVGISIMTYGQHF-UHFFFAOYSA-N
SMILESN.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
pH4.0 to 5.0 (10 % in water)
UVat 280 nm A: =<0.05
Water=<1.0 %
UVat 260 nm A: =<0.05
UV(10 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only