Ion PGM™ 200 Sequencing Kit - FAQs
Ion PGM™ 200 Sequencing Kit - FAQs
View additional product information for Ion PGM™ 200 Sequencing Kit - FAQs (4474004)
1 product FAQs found
How long can I wait until I start the run after initialization of my Ion PGM System?
If you are using the Ion PGM™ 200 Sequencing Kit v2, you can perform a run within 27 hours after initialization. If you are going to perform two runs, make sure the second one starts within 27 hours.
If you are using the Ion PGM™ 200 Sequencing Kit, the limit is shorter; you will need to perform a run within 10 hours.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.