Sodium perchlorate, ACS reagent, anhydrous
Sodium perchlorate, ACS reagent, anhydrous
Sodium perchlorate, ACS reagent, anhydrous
Thermo Scientific Chemicals

Sodium perchlorate, ACS reagent, anhydrous

CAS: 7601-89-0 ∣ ClNaO4 ∣ 122.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 447420025
Price (EUR)
503,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
503,00
Each
Chemical Identifiers
CAS7601-89-0
IUPAC Namesodium perchlorate
Molecular FormulaClNaO4
InChI KeyBAZAXWOYCMUHIX-UHFFFAOYSA-M
SMILES[Na+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric98.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only