Need help deciding what product to use with the Ion PGM™ System? Check out our helpful Applications Decision Tree for the Ion PGM™ System.
Ion Xpress™ Barcode Adapters 81-96 Kit
Ion Torrent™

Ion Xpress™ Barcode Adapters 81-96 Kit

The Ion Xpress™ Barcode Adapters 81-96 Kit provides a set of 16 unique barcode adaptors specifically designed and validated for optimal performance with the Ion Torrent PGM™ and Proton™ semiconductor sequencers.
Have Questions?
Catalog NumberQuantity
44745211 kit
Catalog number 4474521
Price (USD)
3,155.00
Each
Quantity:
1 kit
Price (USD)
3,155.00
Each

The Ion Xpress™ Barcode Adapters 81-96 Kit provides a set of 16 unique barcode adaptors specifically designed and validated for optimal performance with the Ion Torrent PGM™ and Proton™ semiconductor sequencers. When used in combination with the Ion Xpress™ Plus Fragment Library Kit or the Ion Plus Fragment Library Kit, this kit enables users to pool up to 16 fragment libraries prior to emulsion PCR and then conduct multiplexed sequencing analysis, simplifying the Ion semiconductor sequencing workflow for a wide range of applications, including targeted enrichment. Use of this kit with other Barcode Adapters kits allows pooling of up to 96 fragment libraries.

Features of the Ion Xpress™ Barcode Adapters 81-96 Kit include:

  • Enables multiplexing of numerous fragment library samples on a single sequencing chip with the use of robust molecular barcodes
  • Both sequence- and flow-optimized for equal representation all barcodes in a pool and more economical multiplexed sequencing runs
  • Robust error correction built in by design for added confidence in sample identification

Multiplexing with Barcode Adaptors Enables Higher Throughput
Multiplexing with barcoded libraries leads to more cost effective runs by permitting up to 96 samples per run (16 with this kit), significantly decreasing the cost and handling requirements of emPCR, enrichment, and sequencing.

Sequence- and Flow-Optimized for Increased Performance and Efficiency
Barcode adapters contain sequences that are optimized to provide equal representation of all barcodes in a pool, require a minimum number of flows to interrogate, and require only 17 bases of adaptor sequencing required which results in more economical multiplexed sequencing runs.

Robust Error Correction Built In by Design
Built-in error correction virtually eliminates the possibility of incorrect sample assignment.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)Ion GeneStudio™ S5 Series Sequencer, Ion Chef™ System
LibrariesFragment Library
Product TypeBarcode Adapter Kit
Quantity1 kit
Sequencing TypeGenome & DNA Sequencing
Shipping ConditionDry Ice
Throughput96 Samples per Run
Workflow StepLibrary Barcoding
Product LineIon Torrent
Starting MaterialDNA
Unit SizeEach
Contents & Storage
1 box of 17 tubes, store -20°C

Frequently asked questions (FAQs)

Do libraries constructed with the Ion Xpress Barcode adapters require balancing to help ensure equal representation on the Ion PGM or Proton instrument sequencing run?

No, the Ion Xpress Barcode adapters are designed with a 3 bp adapter sequence (GAT) at the 3' end of each barcode to help minimize adapter ligation bias. When pooling barcoded libraries, accurate library quantification is extremely important to help ensure even barcode representation on the sequencing run.

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

How many samples can I multiplex on a single chip for sequencing on the Ion PGM or Proton System?

The number of samples that can be multiplexed in a single sequencing run depends on the capacity of the chip, the size of the library, and the required coverage. A table of approximate capacities based on the size of the library and chip can be found in the Ion Ampliseq Preparation User Guide (https://tools.thermofisher.com/content/sfs/manuals/MAN0006735_AmpliSeq_DNA_RNA_LibPrep_UG.pdf) within the section “Strategies for combining Ion Ampliseq libraries.”

Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.

I am planning to use the Oncomine BRCA Research Assay, Manual Library Preparation (Cat. No. A32840). Which other products would need I to order?

Here are the products you would need to order:

- Ion Xpress Barcode Adapters Kits
- Agencourt AMPure XP (Beckman Coulter Cat. No. A63880 or A63881)
- Ion Library Equalizer Kit (Cat. No. 4482298)