tert-Butyl methyl ether, ACS reagent
Thermo Scientific Chemicals

tert-Butyl methyl ether, ACS reagent

CAS: 1634-04-4 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number 447470010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceClear liquid
Color scale=<10 APHA
GC>=99.0 %
Residue after evaporation=<0.001 %
Water=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only