2-Methoxyethanol, ACS reagent
2-Methoxyethanol, ACS reagent
2-Methoxyethanol, ACS reagent
Thermo Scientific Chemicals

2-Methoxyethanol, ACS reagent

CAS: 109-86-4 | C3H8O2 | 76.09 g/mol
Catalog NumberQuantity
4474800101 L
Catalog number 447480010
Price (JPY)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.3 %
Water=<0.1 %
Titrable acid=<0.002 meq/g
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only