Calcium nitrate tetrahydrate, 98%, extra pure
Calcium nitrate tetrahydrate, 98%, extra pure
Calcium nitrate tetrahydrate, 98%, extra pure
Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, 98%, extra pure

CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Catalog number 447530050
Price (EUR)
112,00
Each
Quantity:
5 kg
Request bulk or custom format
Price (EUR)
112,00
Each
Chemical Identifiers
CAS13477-34-4
Molecular FormulaCaH8N2O10
InChI KeyMWGCGFYACDTFSB-UHFFFAOYSA-N
SMILESO.[Ca++].[O-][N+]([O-])=O
Molecular Weight (g/mol)236.15
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric97.5 to 102.5 %
Solubility(20 g + 100 ml water) Clear colorless solution
Appearance (Form)Crystals
pH5.0 to 9.0 (5 % at 25°C)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only