Where can I find the sequences for the Ion Xpress RNA-Seq Barcodes 1-16 (Cat. No. 4475485)?
The Ion Xpress RNA-Seq Barcode 1-16 sequences are the same as the DNA library Ion Xpress Barcode Adapter 1-16 sequences and can be found under References in the Torrent Browser.