Magnetic Bead Purification Module
Ion Torrent™

Magnetic Bead Purification Module

The Magnetic Bead Purification Module is a cleanup kit used in the construction of RNA libraries for sequencing on the Ion Torrent PGM™ sequencer.
Have Questions?
Catalog NumberQuantity
447548612 Preps
Catalog number 4475486
Price (USD)
406.00
Each
Quantity:
12 Preps
Price (USD)
406.00
Each

The Magnetic Bead Purification Module is a cleanup kit used in the construction of RNA libraries for sequencing on the Ion Torrent PGM™ sequencer. This module is included in the Ion Total RNA-Seq Kit v2, but may also be purchased separately if additional modules are needed.

For small RNA library construction using the Ion Total RNA-Seq Kit v2, the module is used for cleanup after cDNA synthesis, PCR amplification, and for the small RNA enrichment steps. This magnetic bead-based cDNA cleanup step replaces the gel size selection step in the previous version of the kit. For whole transcriptome library construction using the Ion Total RNA-Seq Kit v2, the module is used for cleanup after the fragmentation, cDNA, and PCR amplification steps.

This module is not for use alone or in conjuction with any kits other than the Ion Total RNA-Seq Kit v2.

Related Links:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Sequencing
No. of Reactions12 Preps
Product TypeMagnetic Bead Purification Module
Quantity12 Preps
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
1 Processing Plate — Store at Room Temperature
1 x 11 ml Bottle Binding Solution Concentrate — Store at Room Temperature
1 x 11 ml BottleWash Solution Concentrate — Store at Room Temperature
1 x 0.7 ml Tube Nucleic Acid Binding Beads — Store at 4 °C *DO NOT FREEZE*