Product Image
Applied Biosystems™

5500 W Buffer G

5500 W Buffer G is needed for ligation sequencing on the 5500 W Series genetic analyzers.
Have Questions?
Catalog NumberQuantity
44756221 kit
Catalog number 4475622
Price (USD)
-
Quantity:
1 kit

5500 W Buffer G is needed for ligation sequencing on the 5500 W Series genetic analyzers. Supplemental buffers like this one can be used to replace components of the main buffer kit. 5500 W Buffer G contains sufficient reagents to run up to 432 cycles, or 72 cycles per lane for 6 lanes.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
BufferSequencing Buffer
For Use With (Equipment)5500 Genetic Analyzer, 5500xl Genetic Analyzer
Product Type5500 W Buffer G
Quantity1 kit
Throughput432 Cycles per Lane, 72 Cycles per Lane
Product LineSOLiD
Unit Size1 kit
Contents & Storage
Store at ambient temperature