Fosfato de potasio, dibásico, trihidrato, extra puro, Thermo Scientific Chemicals
Fosfato de potasio, dibásico, trihidrato, extra puro, Thermo Scientific Chemicals
Fosfato de potasio, dibásico, trihidrato, extra puro, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fosfato de potasio, dibásico, trihidrato, extra puro, Thermo Scientific Chemicals

CAS: 16788-57-1 | H7K2O7P | 228.22 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 g
1 kg
Envase:
Frasco de plástico
Número de catálogo 447730010
Precio (EUR)
148,00
Each
Cantidad:
1 kg
Envase:
Frasco de plástico
Pedido a granel o personalizado
Precio (EUR)
148,00
Each
Identificadores químicos
CAS16788-57-1
IUPAC Namedipotassium trihydrate hydrogen phosphate
Molecular FormulaH7K2O7P
InChI KeyXQGPKZUNMMFTAL-UHFFFAOYSA-L
SMILESO.O.O.[K+].[K+].OP([O-])([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White
Sodium (Na)=<0.1 %
Appearance (Form)Crystals
Titration with HClO4>=98.5 %
pH8.5 to 9.6 (5 % solution)
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only