Ion Xpress™ Plus Fragment Library Kit for AB Library Builder™ System - FAQs
Ion Xpress™ Plus Fragment Library Kit for AB Library Builder™ System - FAQs
View additional product information for Ion Xpress™ Plus Fragment Library Kit for AB Library Builder™ System - FAQs (4477598)
2 product FAQs found
Are there Ion Torrent fragment library kits available for use with my AB Library Builder System?
The currently available kits for the AB Library Builder System instrument includes:
- Ion Plus Library Kit for AB Library Builder System (Cat. No. 4477597)
- Ion Xpress Plus Library Kit for AB Library Builder System (Cat. No. 4477598).
Both kits require the use of the separately available Ion Plus Fragment Library Adapters Kit (Cat. No. 4476340) or the Ion Xpress Barcode Adapters kits and the Ion Plus and Ion Xpress Plus Protocol Card for AB Library Builder System (Cat. No. 4477687).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many samples can I multiplex on a single chip for sequencing on the Ion PGM or Proton System?
The number of samples that can be multiplexed in a single sequencing run depends on the capacity of the chip, the size of the library, and the required coverage. A table of approximate capacities based on the size of the library and chip can be found in the Ion Ampliseq Preparation User Guide (https://tools.thermofisher.com/content/sfs/manuals/MAN0006735_AmpliSeq_DNA_RNA_LibPrep_UG.pdf) within the section “Strategies for combining Ion Ampliseq libraries.”
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.