Tau Polyclonal Antibody
Invitrogen™

Tau Polyclonal Antibody

This rabbit anti-Tau polyclonal antibody kit is specific to TAU and is reactive to human. Rabbit anti-Tau polyclonal antibody kitMás información
Have Questions?
Número de catálogoConjugado
44779G
también denominado 44-779G
Unconjugated
Número de catálogo 44779G
también denominado 44-779G
Precio (MXN)
-
Conjugado:
Unconjugated
This rabbit anti-Tau polyclonal antibody kit is specific to TAU and is reactive to human. Rabbit anti-Tau polyclonal antibody kit recognizes the expressed product of the MAPT gene. Tau is a neuronal microtubule-associated protein found predominantly on axons and functions to promote tubulin polymerization and stabilize microtubules. Validated applications for rabbit anti-TAU polyclonal antibody kit are Western blotting, immunoprecipitation and immunostaining.



Applications: Validated applications for rabbit anti-Tau polyclonal antibody kit are Western blotting, immunoprecipitation and Immunostaining.

Host species and isotype: The host species and isotype of the Tau polyclonal antibody kit is rabbit IgG.

Reactivity: Rabbit anti-Tau polyclonal antibody kit detects human TAU.

Product size: Rabbit anti-Tau polyclonal antibody kit is available in 1 kit pack size.



Tau is a neuronal microtubule-associated protein found predominantly on axons and functions to promote tubulin polymerization and stabilize microtubules. Tau, in its hyperphosphorylated form, is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's disease (AD) brain. Hyperphosphorylated Tau is also found in neurofibrillary lesions in a range of other central nervous system disorders. Hyperphosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases including GSK-3β, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II (CK2), phosphorylate Tau. The Tau PSSA Sampler Pack allows one to readily study selective phosphoregulation of multiple sites along Tau. This can be extremely useful in kinase-directed experiments as well as Tau regulation studies.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
ID de gen (Entrez)4137
Host abrvRb
ReactividadHuman
Reactividad abrvHu
Condiciones de envíoApproved for shipment on Wet or Dry Ice
Gen dianaTau
Estado de modificación de destinoPhospho
Abrv de aplicación validadaWB
AplicacionesWestern Blot
ClasificaciónPolyclonal
ConjugadoUnconjugated
Especie del huéspedRabbit
Unit Size1 kit
Contenido y almacenamiento
Contains 11 vials.
Store in freezer (-20°C).
For shipment or short-term storage (up to 1 week), 2–8°C is sufficient.