Sodium azide, 99+%, for biochemistry
Sodium azide, 99+%, for biochemistry
Sodium azide, 99+%, for biochemistry
Thermo Scientific Chemicals

Sodium azide, 99+%, for biochemistry

Sodium azide, CAS # 26628-22-8, is the sodium salt of hydrogen azide (hydrazoic acid). This compound shows antibacterial and mutagenic activities. It also can inhibit the mitochondrial respiratory chain. | CAS: 26628-22-8 | N3Na | 65.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 447815000
Price (USD)
419.65
Online Exclusive
466.00
Save 46.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
419.65
Online Exclusive
466.00
Save 46.35
Each
Chemical Identifiers
CAS26628-22-8
IUPAC Namesodium azide
Molecular FormulaN3Na
InChI KeyPXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES[Na+].[N-]=[N+]=[N-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Argentometric99.0 to 101.0 %
Appearance (Color)White to almost white
Insoluble matter=<0.05 % (in water)
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Sodium azide is a precursor for preparing inorganic azides such as lead azide and silver azide
• One mechanism of action associated with this compound can be related to the inhibition of mitochondrial ATPase, that consequently suppresses oxidative phosphorylation

Applications

• Sodium azide is an inorganic compound with antibacterial and mutagenic activities mainly used in biological samples
• Sodium azide inhibits cytochrome oxidase in gram-negative bacteria. However, its antibacterial capacity is more limited in some gram-positive bacteria (streptococci, pneumococci, lactobacilli), presenting intrinsic resistance
• Sodium azide is used for the introduction of an azide group to prepare a desired compound
• It is useful in biochemical approaches as a probe reagent and a preservative
• This compound can be used to synthetize blue fluorescent copolymers and metal phosphonates

RUO – Research Use Only

General References:

  1. Abdollahi-Alibeik, M.; Moaddeli, A. Multi-component one-pot reaction of aldehyde, hydroxylamine and sodium azide catalyzed by Cu-MCM-41 nanoparticles: a novel method for the synthesis of 5-substituted 1 H-tetrazole derivatives. New J. Chem. 2015, 39 (3), 2116-2122.
  2. Zeng, M.; Yang, Y. H.; Li, J. J.; Chen, Y.; Cui, D. M.; Zhang, C. Copper Catalyzed Synthesis of Aryl Azides from Aryl Bromides and Sodium Azide. Asian J. Chem. 2015, 27 (5), 1698-1700.