What loci does the VeriFiler Direct PCR Amplification Kit target?
The kit targets the following 9 STR loci: TH01, D2S1338, D19S433, D1S1656, D2S441, D10S1248, D12S391, D22S1045.
Search
View additional product information for VeriFiler™ Direct PCR Amplification Kit - FAQs (4479567)
1 product FAQs found