Ion PGM™ Template OT2 400 Kit - FAQs
Ion PGM™ Template OT2 400 Kit - FAQs
View additional product information for Ion PGM™ Template OT2 400 Kit - FAQs (4479878)
12 product FAQs found
What should I do if the centrifuge is still running even though the time remaining is 0:00 at the final spin of the Ion OneTouch 2?
The "time remaining" is not entirely accurate. The centrifuge may keep rotating for about 15 minutes after the time has become 0:00. If it rotates for more than 15 minutes, please contact Technical Support.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What should I do if an "Over Pressure" error appears after I start an Ion OneTouch 2 run?
The pressure inside the instrument may be above normal. If the Ion OneTouch 2 firmware is older than v540, please update it to the latest version.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Can I use the same oil between the Ion PGM OT2 200 Kit and the Ion PGM OT2 400 Kit?
No, you may not. Please exchange the oil when switching between different kits. Contact Technical Support for any needed assistance when switching kits.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are these black precipitates in the recovery tube and waste of the Ion OneTouch System?
It may be rubber from the packing. The amount varies among instruments and it is not anything to be concerned about, as it will be removed by the Ion OneTouch ES.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How do I update my Ion OneTouch 2 firmware?
Firmware updates to the software controlling the Ion OneTouch 2 Instrument are periodically released. To update the firmware to the appropriate version, use either a USB flash drive or an ethernet connection, as described on page 11 of the Ion OneTouch 2 System - Installation, Setup, and Maintenance User Guide (http://tools.thermofisher.com/content/sfs/manuals/MAN0014388_IonOneTouch2Sys_UG.pdf ).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
My Ion OneTouch 2 run is finished, but I cannot perform enrichment right now. How should I store my unenriched ISPs?
We recommend removing the samples from the Ion OneTouch 2 instrument ≤ 16 hours after starting the run. After centrifuging and removing the samples from the instrument, follow the ISP collection protocol to recommended stopping points and follow the instructions for preparing the unenriched ISPs for storage. The ISP suspension can be stored at 2 degrees C to 8 degrees C for up to 3 days.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How much oil and recovery solution is typically consumed during a normal Ion OneTouch 2 run and during a cleaning?
In a typical Ion OneTouch 2 run, you should expect ~18 mm drop in the level of oil in the tube (~22 mL consumed), and ~13 mm drop in the recovery solution (~15 mL consumed). During cleaning, you should expect to collect ~10 mL of oil in the 50 mL conical waste collection tube.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What procedure do I follow when switching between Ion OneTouch 2 template kit types?
Due to differences in reagent formulations between kit types, we recommend following the Demonstrated Protocol: How to Set Up the Ion OneTouch 2 Instrument When Switching Between the Ion PGM System and Ion Proton System (http://ioncommunity.thermofisher.com/docs/DOC-7265) if using more than one Ion OneTouch 2 Template kit type on your instrument. Briefly, the instrument lines must be primed with the new kit reagents by running a cleaning and initialization procedure prior to setting up the next run. Please see the above user guide for detailed instructions.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How much library do I need as input into the Ion OneTouch 2 template preparation reaction?
The input amount for template preparation on the Ion OneTouch 2 is library type specific. Occasionally, library input titration may be required to optimize templated ISP clonality and maximize sequencing throughput. Please see the specific guidelines for your library type in the appropriate library preparation user guide that can be found here: http://www.thermofisher.com/us/en/home/technical-resources/technical-reference-library/next-generation-sequencing-support-center/ngs-library-preparation-support.html
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
What are the currently available Ion OneTouch 2 template kits?
For the Ion PGM System, we offer:
- Ion PGM Hi-Q View OT2 Kit (Cat. No. A29900) for template preparation of libraries up to 400 bp in length.
For the Ion Proton System, we offer:
- Ion PI Hi-Q OT2 200 Kit (Cat. No. A26434) for template preparation of libraries up to 200 bp in length.
For the Ion S5 System and Ion S5 XL System, we offer two kits:
- Ion 520/530 Kit-OT2 (Cat. No. A27751) for template preparation of libraries up to 400 bp in length.
- Ion 540 Kit- OT2 (Cat. No. A27753) for template preparation of libraries up to 200 bp in length.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How much oil and recovery solution is typically consumed during a normal Ion Torrent Ion OneTouch 2 run and during a cleaning?
In a typical Ion OneTouch 2 run, you should expect ~18 mm drop in the level of oil in the tube (~22 mL consumed), and ~13 mm drop in the recovery solution (~15 mL consumed). During cleaning, you should expect to collect ~10 mL of oil in the 50 mL conical waste collection tube.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
Where can I find the protocols for my Ion Torrent Ion OneTouch 2 kit?
Ion Torrent OneTouch2 protocols can be found at our NGS Support Center (thermofisher.com/ngssupport), in the NGS Template Preparation Support Center section. For all other protocols, please go here (http://www.thermofisher.com/manuals-protocols-search.html).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.