Where can I find the protocols for Ion library preparation?
You can find library protocols, as well as other valuable information on NGS Sequencing, at our NGS Support Center (thermofisher.com/ngssupport). For all other protocols, please go here (http://www.thermofisher.com/manuals-protocols-search.html).