Lysis Buffer, FS
Applied Biosystems™
Lysis Buffer, FS
Lysis Buffer, FS is designed specifically to lyse microbial organisms from food and environmental samples and prepare the microbial DNA for subsequent downstream analysis by real-time PCR.
Have Questions?
Catalog NumberQuantity
44807247 mL
Catalog number 4480724
Price (USD)
-
Quantity:
7 mL
Lysis Buffer, FS is designed specifically to lyse microbial organisms from food and environmental samples and prepare the microbial DNA for subsequent downstream analysis by real-time PCR.
This lysis buffer can be used in conjunction with Proteinase K, FS in the Pathatrix™ Auto Instrument or RapidFinder™ Direct Lysis workflows.
When used as a part of the Pathatrix™ Auto Instrument workflow, one bottle of Lysis Buffer, FS is sufficient for 700 reactions.
When used as a part of the RapidFinder™ Direct Lysis workflow, one bottle of Lysis Buffer, FS is sufficient for 100 reactions.
Related Links:
Specifications
Quantity7 mL
Shipping ConditionRoom Temperature
FormLiquid
Product LinePathatrix
Unit SizeEach
Contents & Storage
Store at 4°C