Sodium phosphate, dibasic, 98+%, extra pure, anhydrous
Sodium phosphate, dibasic, 98+%, extra pure, anhydrous
Sodium phosphate, dibasic, 98+%, extra pure, anhydrous
Thermo Scientific Chemicals

Sodium phosphate, dibasic, 98+%, extra pure, anhydrous

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
25 kg
Catalog number 448140010
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 30-Jun-2026
164.00
Save 24.35 (15%)
Each
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration with HClO498.0 to 102.0 % (On dried substance)
Loss on drying=<5 % (130°C)
pH8.7 to 9.4 (5 % in water at 25°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only