Nunc™ Microplate Lids, 384 and 1536 plates - FAQs
Nunc™ Microplate Lids, 384 and 1536 plates - FAQs
View additional product information for Nunc™ Microplate Lids - FAQs (448143)
1 product FAQs found
Do you offer Nunc Microplate Lids for the Nunc Black or White 384-Well Microplates?
We offer Nunc Microplate Lids: Cat. No. 448143, that fit both the 384-well microplates and the 1,536-well microplates.