Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ 3 x 32-Well Retainers
Applied Biosystems™

MicroAmp™ 3 x 32-Well Retainers

The Applied Biosystems MicroAmp 3 x 32-Well Retainer is specifically designed for use with single tubes and tube strips onRead more
Have Questions?
Catalog NumberQuantity
44816691 retainer
Catalog number 4481669
Price (USD)
41.77
Each
Quantity:
1 retainer
Price (USD)
41.77
Each
The Applied Biosystems MicroAmp 3 x 32-Well Retainer is specifically designed for use with single tubes and tube strips on the ProFlex 3 x 32-Well PCR System. The retainer provides support and allows pressure to be distributed evenly over tubes and strips to ensure a proper seal and prevent the tubes and strips from being crushed or deformed. Its use is recommended at all times when working with MicroAmp reaction tubes and tube strips. Simply place the retainer on the sample block, then load the tubes or strips.
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Application)PCR
For Use With (Equipment)ProFlex 3 x 32-Well PCR System
Plate Compatibility32-Well Plate
Product TypeRetainer
Quantity1 retainer
Shipping ConditionRoom Temperature
ColorBlack
Product LineMicroAmp
Unit SizeEach
Contents & Storage
Store at room temperature.