Ion PGM™ Controls Kit v2 - FAQs
Ion PGM™ Controls Kit v2 - FAQs
View additional product information for Ion PGM™ Controls Kit v2 - FAQs (4482010)
1 product FAQs found
Where can I find the protocols for Ion library preparation?
You can find library protocols, as well as other valuable information on NGS Sequencing, at our NGS Support Center (thermofisher.com/ngssupport). For all other protocols, please go here (http://www.thermofisher.com/manuals-protocols-search.html).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.