What chips are available for the Ion Proton System and what are the specifications?
Currently, the Ion Proton System supports the Ion PI Chip Kit v2 (Cat. No. 4482321), which contains 165 million wells and typically generates up to 10 Gb of data and ~60-80 million reads.