I am new to making RNA libraries using the Ion Total RNA-Seq Kit v2 (Cat. No. 4475936). What is a good approach?
Please visit the RNA Sequencing applications (https://www.thermofisher.com/us/en/home/life-science/sequencing/rna-sequencing.html) for information regarding sequencing data analysis, including white papers, training videos for transcriptome data analysis, and information about third-party software solutions (i.e., Partek Inc.).