For licensing and limited use restrictions visit thermofisher.com/HIDlicensing.
Yfiler™ Plus PCR Amplification Kit
Applied Biosystems™

Yfiler™ Plus PCR Amplification Kit

Note: The Yfiler Plus kit is now approved for use by laboratories generating DNA profiles for inclusion in the USRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4482730500 Reactions
4484678100 Reactions
Catalog number 4482730
Price (USD)
26,160.00
Each
No. of Reactions:
500 Reactions
Price (USD)
26,160.00
Each
Note: The Yfiler Plus kit is now approved for use by laboratories generating DNA profiles for inclusion in the US National DNA Index System (NDIS) CODIS database.

The Yfiler Plus PCR Amplification Kit is designed for forensic laboratories that process highly challenging Y-STR casework and database samples. The Yfiler Plus kit is optimized to help improve resolution of trace evidence and sexual assault mixtures that contain small quantities of male DNA in a large background of female DNA and offers laboratories both the power of discrimination and the performance to enable accurate results in less time from challenging samples.

The Yfiler Plus PCR Amplification Kit is a 6-dye multiplex assay for short tandem repeats (STRs) that allows amplification from multiple male-specific sample types, such as male–male and male–female mixtures, and direct PCR amplification from single-source samples.

Key features of the Yfiler Plus PCR Amplification Kit:
• Improved discrimination capacity through new, highly discriminating markers
• Improved performance with degraded samples and PCR inhibitors
• Improved genotyping accuracy through expanded allelic ladder and virtual bins
• Concordance with existing data in reference haplotype databases
• Increased workflow efficiency enabling faster time-to-results than previous Y-STR kits

Improved discrimination capacity
The Yfiler Plus PCR Amplification Kit enables multiplex amplification of the 17 Y-STR markers included in the AmpFLSTR Yfiler PCR Amplification Kits, as well as 10 new Y-STRs using 6-dye chemistry to offer the highest discriminatory capacity of any commercially available Y-STR kit. In particular, the inclusion of seven rapidly-mutating (RM) Y-STRs with mutation rates above 1 × 10(-2) helps improve resolution of paternal lineage differentiation as well as helps with discrimination of closely-related males.

More information from challenging casework samples
Y-STR analysis is useful for processing of complex casework samples with trace quantities of male DNA in presence of high level of female DNA. These samples may include rape kit samples, body fluid with few or no sperm present, and fingernail scrapings from female victims of violent assaults. These samples often show DNA degradation and contain inhibitors that can make the analysis difficult and lead to inconclusive results. The Yfiler Plus kit harnesses the latest developments in PCR amplification technology to help provide cleaner baseline and improved recovery of alleles with mixtures and highly inhibited samples.

Streamlined protocol and increased workflow efficiency
The Yfiler Plus kit provides faster thermal cycling conditions enabling shorter time to result. The kit is also designed to process single-source reference samples using direct PCR amplification from the following types of samples:
• Blood and buccal samples on treated paper substrates, without the need for sample purification
• Blood samples collected on untreated paper substrates and treated with Prep-n-Go Buffer
• Buccal samples collected on swab substrates and treated with Prep-n-Go Buffer

Complete validated solution
Our products provide a complete solution for your human identification needs and include DNA quantification kits, a range of STR kits, instrumentation, software, and consumables. The Yfiler Plus kit is part of a comprehensively validated system for STR-based human identification. All reagents are developed in accordance with guidelines from leading worldwide forensic organizations such as SWGDAM and DAB.

For licensing and limited use restrictions visit thermofisher.com/HIDlicensing.
Specifications
For Use With (Application)Sequencing
Label or Dye6-FAM, LIZ, NED, SID, TAZ, VIC
No. of Reactions500 Reactions
Product LineYfiler
Product TypePlus PCR Amplification Kit
Quantity500 reactions
Shipping ConditionDry Ice
Target LociDYS438, DYS627, DYS458, DYS437, DYS391, DYS392, DYS635 (Y GATA C4), DYS19, DYS390, DYS439, DYS456, DYS393, DYS449, DYS387S1 a⁄b, DYS576, DYS460, DYS533, DYS389 I⁄II, DYS570, DYS385 a⁄b, DYS481, YGATA H4, DYS518, DYS448
FormatKit
Unit SizeEach
Contents & Storage
• Yfiler Plus Master Mix, 4 tubes, 0.5 mL/tube
• DNA Control 007, 2 tubes, 0.05 mL
• Yfiler Plus Primer Set, 2 tubes, 0.25 mL/tube, protect from light
• Yfiler Plus Allelic Ladder, 2 tubes, 0.025 mL/tube, protect from light

Store at –5 to –30°C upon receipt and at 2–8°C after initial use.

Frequently asked questions (FAQs)

How can knowledge of the ratio of male to female DNA present in a mixed sample be used to determine the best HID analysis kit for the sample?

Knowing the ratio of male to female DNA present in a mixed sample can help the analyst to decide whether autosomal or Y-STR analysis will provide the maximum amount of information on the male component. Generally, for samples with a male:female DNA ratio up to 1:10, both male and female components can usually be visualized and interpreted using an autosomal STR system such as the Quantifiler Duo DNA Quantification Kit. For samples with a male:female DNA ratio above 1:10, the minor component may be below the level of detection of the instrument platform if analyzed using an autosomal STR kit, and therefore the male component may best be analyzed using a Y-STR system such as the Yfiler Plus kit. Each laboratory should determine the minimum level of detection for the male component of a mixed sample using an autosomal amplification system and identify the cut-off value at which Y-STR analysis may prove more successful. This can be achieved as part of an internal validation study of the Quantifiler Duo DNA Quantification Kit.