3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98%, Thermo Scientific Chemicals
3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98%, Thermo Scientific Chemicals
3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3',5,5'-Tetra-tert-butyldiphenoquinone, 98%, Thermo Scientific Chemicals

CAS: 2455-14-3 | C28H40O2 | 408.63 g/mol
Have Questions?
Catalog NumberQuantity
4483000202 g
Catalog number 448300020
Price (USD)
-
Quantity:
2 g
Request bulk or custom format
Chemical Identifiers
CAS2455-14-3
IUPAC Name3,3',5,5'-tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione
Molecular FormulaC28H40O2
InChI KeyGQIGHOCYKUBBOE-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC(C=C(C1=O)C(C)(C)C)=C1C=C(C(=O)C(=C1)C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown to red
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only