MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates with Barcode
Applied Biosystems™

MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates with Barcode

Applied Biosystems™ MicroAmp™ EnduraPlate™ plastic consumables offer excellent PCR or qPCR performance in formats developed to meet your experimental needs.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
448335420 Plates
4483352500 Plates
카탈로그 번호 4483354
제품 가격(KRW)
266,000
온라인 행사
Ends: 31-Dec-2026
295,000
할인액 29,000 (10%)
Each
수량:
20 Plates
제품 가격(KRW)
266,000
온라인 행사
Ends: 31-Dec-2026
295,000
할인액 29,000 (10%)
Each
Applied Biosystems™ MicroAmp™ EnduraPlate™ plastic consumables offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. All of our plastic consumables are validated with Applied Biosystems™ instruments for optimal fit and performance. With the new EnduraPlate™ reaction plates, there is now a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments.

Key Features and Benefits:

• Optimized for performance with Applied Biosystems™ instruments
• Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
• Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
• Available in multiple colors for easy organization of laboratory assays
• Excellent mechanical stability and flatness for your complete workflow needs

Use of polypropylene and a high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate provides mechanical stability, flatness, and excellent durability. ANSI/SBS compliant construction combined with mechanical durability during and after thermal cycling makes EnduraPlate™ reaction plates an ideal choice for use with automation/robotics and multi-instrument experiments (e.g., thermal cycler -> real-time thermal cycler -> genetic analyzer).

EnduraPlate™ reaction plates have a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from thermal block to sample. These plates are optimized for performance with Applied Biosystems™ instruments. Easy-to-read, black well identification text helps to minimize pipetting mistakes and improves sample preparation productivity, while five color choices provide a convenient way to organize your laboratory assays.

Note: An adapter (Cat No. A24820) is required for use of this product with 7300/7500 Real-Time PCR systems.
사양
인증/적합성Certified DNA/RNase Free
용도(장비)Thermal Cycler, Genetic Analyzer, DNA Analyzer
포장20 Plates
제품 유형Microplate
수량20 Plates
반응 속도Standard
스커트 스타일Semi
부피(미터법) 웰0.3 mL
부피(미터법) 작동0.2 mL
바코드Includes Barcode
색상Clear, Optical
물질Polycarbonate, Polypropylene
웰 수96
제품라인MicroAmp
Unit SizeEach
구성 및 보관
Store at room temperature.

자주 묻는 질문(FAQ)

What is included in Oncomine Precision Assay GX (Cat. No. A46291)?

Oncomine Precision Assay GX (Cat. No. A46291) includes the following:
- OPA Pool 1 panel (Part No. A44350)
- Genexus Strip 1, quantity of 8 strips (Part No. A46812)
- Genexus Strip 2-HD, quantity of 8 strips (Part No. A46814)
- The OPA Pool 1 is provided in 8 pairs of tubes, where each tube pair contains one tube with forward (FWD) primers and one tube with reverse (REV) primers.