MicroAmp™ EnduraPlate™ Optical 96-Well Fast GPLE Clear Reaction Plates with Barcode
Applied Biosystems™ MicroAmp™ EnduraPlate™ GPLE plastic consumables provide superior PCR and qPCR performance and quality in formats developed to meetRead more
Have Questions?
Catalog Number
Quantity
4483481
20 plates
Catalog number 4483481
Price (MXN)
-
Quantity:
20 plates
Applied Biosystems™ MicroAmp™ EnduraPlate™ GPLE plastic consumables provide superior PCR and qPCR performance and quality in formats developed to meet your experimental needs. All EnduraPlate™ GPLE plastic consumables are validated on Applied Biosystems™ thermal cyclers, real-time thermal cyclers, and genetic analyzers for optimal fit and performance.
Key Features and Benefits:
• Each lot is certified DNA, RNAse, and PCR inhibitor-free • Labeled 'For Laboratory Use' • Optimized for performance with Applied Biosystems™ instruments • Excellent mechanical stability and flatness for your complete workflow needs
Use of polypropylene and a high temperature molding process helps prevent DNA, RNA, and enzymes from binding, while polycarbonate offers mechanical stability, flatness, and excellent durability. ANSI/SBS-compliant construction combined with mechanical durability during and after thermal cycling makes EnduraPlate™ reaction plates an ideal choice for use with automation/robotics and multi-instrument experiments (e.g., thermal cycler → real-time thermal cycler → genetic analyzer).
EnduraPlate™ reaction plates have a thin-wall construction that allows for a snug fit to thermal blocks with excellent heat transfer from thermal block to sample. These plates are optimized for performance with Applied Biosystems™ instruments. Easy-to-read, black, well-identification text helps to minimize pipetting mistakes and improves sample preparation productivity.
Note: An adapter (Cat No. A24820) is required for use of this product with 7300/7500 Real-Time PCR systems.
For Laboratory Use.
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer