High-Power USB Wi-Fi Module - FAQs
High-Power USB Wi-Fi Module - FAQs
View additional product information for High-Power USB Wi-Fi Module - FAQs (4483658)
1 product FAQs found
Is there wireless connectivity with the SimpliAmp Thermal Cycler?
Like the ProFlex PCR System, the SimpliAmp Thermal Cycler can be connected wirelessly with an optional High-Power USB W-Fi Module, Cat. No. A26774.