NAOBasket™ + 2 mL Tube, in peel pouch
Thermo Fisher™
NAOBasket™ + 2 mL Tube, in peel pouch
Nucleic Acid Optimizer (NAOBasket) collapsable grid allows lysis in the basket. Elute with high speed centrifugation.
Have Questions?
Número de catálogoIncluye
4483798NAOBasket and 2 mL tube in peel pouch
Número de catálogo 4483798
Precio (MXN)
-
Incluye:
NAOBasket and 2 mL tube in peel pouch
The Nucleic Acid Optimizer basket (NAOBasket) is a plastic semi-permeable device that allows fast and complete recovery of nucleic acids from swab samples, card punches, or other specimens into a sample tube.
NAOBasket is designed with a collapsable grid bottom that allows passage of all eluted DNA from the collection device (i.e., swab) into a sample tube during high-speed centrifugation, with a tight fit to help prevent leaks during vortexing.
Features of NAOBasket include:
  • Forensic DNA grade—free of amplifiable human DNA and detectable DNase and RNase; EtO treated
  • Lyse and centrifuge in the basket—innovative product design allows the passage of DNA from sample into the tube using a grid bottom, and tight leak-proof design can endure vortexing, centrifugations, and various storage conditions
  • Increased DNA yield particularly when combined with FLOQSwabs®
  • Decreased risk of sample contamination—significant reduction of manual manipulation of the sample
  • Compatibility—compatible with AutoMate Express Forensic DNA Extraction System (Cat. No. 4441763), HID Nimbus Presto System (Cat. No. A55395), and HID NIMBUS Presto QNA System (Cat. No. A55769)
NAOBasket is manufactured by Copan, a market leader in the area of sample collection tools, and distributed globally by Thermo Fisher Scientific.
For Criminal Justice and Forensic Use Only.
Especificaciones
ColorTranslucent
IncluyeNAOBasket and 2 mL tube in peel pouch
Tipo de productoNAOBasket
Unit Size100 pcs/pack