Which sequencing platform is the Ion PGM IC 200 Kit compatible with?
The Ion PGM IC 200 Kit is only compatible with the Ion Chef system and the Ion PGM system and should not be replaced or interchanged with other product kit components.