Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?
Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.