4485467 - FAQs
4485467 - FAQs
View additional product information for 4485467 - FAQs (4485467)
3 product FAQs found
What are the storage recommendations for TaqMan Array Cards (TAC)?
We recommend storing TaqMan Array Cards (TAC) at 2-8 degrees C. TaqMan Array Cards (TAC) are stable for 2 years from the manufacturing date. The expiration date is listed on the packaging.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Where can I find a user guide for the Specialty Configurator tool?
The Specialty Configurator tool user guide can be downloaded from the Specialty Configurator Tool landing page (https://specialty-taqman-arrays.com/home.html) or accessed directly using the following link: https://specialty-taqman-arrays.com/content/dam/LifeTech/global/life-sciences/CellCultureandTransfection/platings/pdf/Specialty%20TAP%20TAC%20Configurator%20Tool%20User%20Guide.pdf.
What is the online Specialty Configurator tool, and how can I access it?
The online Specialty Configurator tool allows for the configuration of Specialty TaqMan Array Plates and Cards that cannot be created using the Custom Array Configurator. This includes 384-well TaqMan Array Plates and TaqMan Array Plate/TaqMan Array Card products containing at least one Custom TaqMan assay or the following assay types:
• TaqMan SNP Genotyping assay
• TaqMan Copy Number assay
• TaqMan miRNA assay (Classic or Advanced)
• TaqMan Mutation Detection assay
The Specialty Configurator tool can also be used to obtain price estimates and quotes for ordering. You can access the tool on the Thermo Fisher Scientific website using the following link: https://specialty-taqman-arrays.com