QuantStudio™ 3D Digital PCR Chip Adapter Kit for Flat Block Thermal Cycler - FAQs
QuantStudio™ 3D Digital PCR Chip Adapter Kit for Flat Block Thermal Cycler - FAQs
View additional product information for QuantStudio™ 3D Digital PCR Chip Adapter Kit for Flat Block Thermal Cycler - FAQs (4485513)
26 product FAQs found
How do I figure out the concentration of my DNA stock from the results I get from the QuantStudio 3D Digital PCR System?
If you include all of the necessary dilution factors into the AnalysisSuite software, the software will give you the copies/µL in your stock. There are 2 dilutions you need to take into account. The first is the dilution of the sample in the reaction. The second is the dilution of the stock that you make before adding it to the digital PCR reaction.
For example, say you want to add 1 µL of a sample that has been diluted 1:10 from the stock. If you add 1 µL of your sample to a 16 µL (final volume) reaction, the dilution factor of the sample is 1:16 or 1/16 = 0.0625. Since the stock has also been diluted 1:10 (0.1), you also need to factor this in. The final dilution factor to enter into the software is 0.0625 x 0.1 = 0.00625 (1:160). As you can see below, you can use either annotation to indicate the dilution factor in the AnalysisSuite software. If you enter that value into the “Dilution” column, the software will give you the copies/µL in your starting material (stock).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
I'm having trouble using AnalysisSuite software on Internet Explorer. What is wrong?
AnalysisSuite software is optimized for Google Chrome v4.0 or later. Firefox also works, but Internet Explorer is not recommended.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How do I update the firmware on the QuantStudio 3D Digital PCR System?
Please follow the procedures starting in "Chapter A: Maintenance" of the QuantStudio 3D Digital PCR System User Guide (https://tools.thermofisher.com/content/sfs/manuals/MAN0007720.pdf) to update the instrument firmware.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How do I replace the fuses in the QuantStudio 3D Digital PCR System?
Please follow the procedures described in "Chapter A: Maintenance" of the QuantStudio 3D Digital PCR System User Guide to replace the fuses. Make sure to only replace with those listed and certified fuses of the same type and rating as those currently in the QuantStudio 3D instrument.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How does sample preparation for digital PCR differ from real-time PCR?
The answer is that sample prep for digital PCR is no different from real-time PCR when using the QuantStudio 3D Digital PCR System. If you have samples in your freezer that you've run on real-time PCR, then you're ready to run them with digital PCR.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How many replicate chips do I have to run?
This will depend on the concentration range of the sample used. The QuantStudio 3D Digital PCR System has application- and concentration-specific requirements for technical replicates. At a low concentration, more replicates will be needed to have confidence in the data. Also, due to the nature of the application, replicates would be needed when studying rare mutations.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How do I determine how much sample to use when setting up the digital PCR reaction for the QuantStudio 3D Digital PCR System?
Please see the section on Sample Dilution in Chapter 3 of the QuantStudio 3D Digital PCR System User Guide (http://tools.thermofisher.com/content/sfs/manuals/MAN0007720.pdf) for more details on how to determine the target copy number per genome.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How many samples can be loaded per chip?
The current QuantStudio 3D Digital PCR Chip allows for one sample per chip; however, duplexing allows analysis of two targets per chip.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How do I connect the QuantStudio 3D Digital PCR System to a network?
Please follow the procedures described starting in Appendix B of the QuantStudio 3D Digital PCR System User Guide (http://tools.thermofisher.com/content/sfs/manuals/MAN0007720.pdf) to connect the instrument to a network.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How quickly can the QuantStudio 3D Digital PCR System read a chip?
The QuantStudio 3D Digital PCR System can read the digital chip in less than 1 minute. In fact, if you are running multiple chips, you can run the next chip in the series when the instrument touch screen displays the Analyzing Chip screen.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How soon do I have to read the QuantStudio 3D Digital Chip after cycling is complete?
After thermal cycling, the QuantStudio 3D Digital PCR 20K Chips can remain on the sample block for up to 24 hours. Read the QuantStudio 3D Digital PCR 20K Chips within 1 hour after you remove them from the GeneAmp 9700 PCR System sample block.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How many QuantStudio 3D Digital PCR Chips can be cycled at one time on the GeneAmp 9700 PCR System?
Using the Chip Carrier on the GeneAmp 9700 Dual Flat Block, up to 24 QuantStudio 3D Digital Chips can be run at one time. Please refer to Chapter 3 in the QuantStudio 3D Digital PCR System User Guide (http://tools.thermofisher.com/content/sfs/manuals/MAN0007720.pdf) for more details on loading the chips for thermal cycling.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Are there any special precautions for loading the digital chips?
Yes. Never handle QuantStudio 3D Digital PCR 20K Chips or Chip Case Lids without gloves. Oils from your hands can contaminate the components and interfere with thermal cycling and imaging. For more instructions, please see the guidelines in Chapter 2 in the QuantStudio 3D Digital PCR System User Guide (http://tools.thermofisher.com/content/sfs/manuals/MAN0007720.pdf) on loading the digital chips.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Does the GeneAmp 9700 PCR System need a particular version of firmware to be compatible with the QuantStudio 3D Digital Chips?
Yes, The GeneAmp 9700 PCR System must be running firmware version 3.12 or later to thermal-cycle QuantStudio 3D Digital PCR 20K Chips. The firmware version of your GeneAmp 9700 PCR System is shown on the main display after you first turn on the instrument. If needed, download and upgrade the firmware.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Does the QuantStudio 3D instrument perform the cycling step, or is that separate?
The QuantStudio 3D instrument only performs the imaging and primary analysis of the digital chips. The chips themselves must be cycled offline on a Dual Flat Block GeneAmp 9700 PCR System.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What thermal cycler can I use to run the QuantStudio 3D Digital PCR Chips?
The QuantStudio 3D Digital PCR Chips are meant to be used on a Dual Flat Block Proflex or GeneAmp 9700 PCR System.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How long is the UV glue good for?
The UV glue is usable for 3 months after first opening, when stored properly. Keep it stored in an amber bag while using it on the bench, and in an amber bag in a dark place between uses.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Where are my chip image data saved on the QuantStudio 3D instrument?
You can configure the QuantStudio 3D instrument to upload the imaging data to a USB drive, a network file server, or the cloud data storage service. See Appendix B, “Networking”, in the QuantStudio 3D Digital PCR System User Guide for more information on setting up network destinations. Also, the QuantStudio 3D instrument retains a copy of the analyzed data from the imaged QuantStudio 3D Digital PCR 20K Chip that you can access from the Run History screen. Note: The results will remain in the QuantStudio 3D instrument cache for approximately 300 readings. After 300 chips, the instrument removes the oldest data file in the cache to store each new reading.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What is the Tilt Base for?
The Tilt Base inclines the GeneAmp 9700 PCR System sample block by 11 degrees C, the angle required for optimal performance of the QuantStudio 3D Digital PCR 20K Chips.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
How long is the immersion fluid good for once the syringe opened?
Use all of the immersion fluid within 60 minutes of uncapping the syringe. Once a syringe is opened, you cannot reattach the cap for later use.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What dyes are supported by the QuantStudio 3D Digital PCR System?
The QuantStudio 3D Digital PCR System features a filter set that is optimized for the FAM, VIC, and ROX dyes available from Life Technologies.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What does the QuantStudio 3D Digital PCR Chip look like?
The QuantStudio 3D Digital PCR 20K Chip is a 10 mm2 high-density reaction plate that consists of a single array of 20,000 reaction wells, where each well can accommodate an 865 pL reaction. Hydrophobic coatings on the chip surface enable loading and isolation of independent PCR reactions within the reaction wells. Please see an image of the chip in the QuantStudio 3D Digital PCR System User Guide, found here: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0007720.pdf
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What are the electrical and environmental specifications of the QuantStudio 3D Digital PCR System?
For instrument specifications, please see Appendix D, “Specifications and Layout”, in the QuantStudio 3D Digital PCR System User Guide for more details at http://tools.thermofisher.com/content/sfs/manuals/MAN0007720.pdf.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What applications can I perform using digital PCR?
Digital PCR can be used to perform rare allele detection, absolute quantitation of gene expression, viral titering, and absolute quantitation of next-generation sequencing libraries.
What systems can be used for digital PCR?
The QuantStudio 12K Flex system with OpenArray block and the QuantStudio 3D system are designed for digital PCR applications.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.