Phospho-ITGB1 (Ser785) Polyclonal Antibody - FAQs
Phospho-ITGB1 (Ser785) Polyclonal Antibody - FAQs
View additional product information for Phospho-ITGB1 (Ser785) Polyclonal Antibody - FAQs (44870G)
1 product FAQs found
Do you offer antibodies for Integrin beta1?
Yes, we have several antibodies against integrin beta 1, including MA513658, PA529606, antibodies against phosphorylated integrin beta 1 phosphorylated at ser785 (Cat. No. 44-870G), as well as anti-integrin beta 1 phosphorylated at Thr788 and789 (Cat. No. 44-872).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.