Immobilized Ficin (Agarose Resin) - FAQs
Immobilized Ficin (Agarose Resin) - FAQs
View additional product information for Immobilized Ficin (Agarose Resin) - FAQs (44881)
1 product FAQs found
Can I purchase the immobilized enzymes (proteases) from the Thermo Scientific IgG Fragmentation kit separately?
Yes. The three immobilized enzymes are available in 5 mL package sizes as shown below:
• Immobilized Ficin, Cat. No. 44881
• Immobilized Papain, Cat. No. 20341
• Immobilized Pepsin, Cat. No. 20343
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.