4,4-Dimethylcyclohexanone, 97%
4,4-Dimethylcyclohexanone, 97%
4,4-Dimethylcyclohexanone, 97%
Thermo Scientific Chemicals

4,4-Dimethylcyclohexanone, 97%

CAS: 4255-62-3 | C8H14O | 126.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
View all
Glass Bottle
Catalog number 448810050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS4255-62-3
IUPAC Name4,4-dimethylcyclohexan-1-one
Molecular FormulaC8H14O
InChI KeyPXQMSTLNSHMSJB-UHFFFAOYSA-N
SMILESCC1(C)CCC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point41°C to 45°C
Appearance (Color)White to tan
Appearance (Form)Crystals or solid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only