4'-Isobutylacetophenone, 97%
4'-Isobutylacetophenone, 97%
4'-Isobutylacetophenone, 97%
Thermo Scientific Chemicals

4'-Isobutylacetophenone, 97%

CAS: 38861-78-8 | C12H16O | 176.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 448822500
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS38861-78-8
IUPAC Name1-[4-(2-methylpropyl)phenyl]ethan-1-one
Molecular FormulaC12H16O
InChI KeyKEAGRYYGYWZVPC-UHFFFAOYSA-N
SMILESCC(C)CC1=CC=C(C=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5170 to 1.5190 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only