5-Methoxyisatin, 97%
5-Methoxyisatin, 97%
5-Methoxyisatin, 97%
Thermo Scientific Chemicals

5-Methoxyisatin, 97%

CAS: 39755-95-8 | C9H7NO3 | 177.16 g/mol
製品番号(カタログ番号) 448830050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS39755-95-8
IUPAC Name5-methoxy-2,3-dihydro-1H-indole-2,3-dione
Molecular FormulaC9H7NO3
InChI KeyDMHGXMPXHPOXBF-UHFFFAOYSA-N
SMILESCOC1=CC=C2NC(=O)C(=O)C2=C1
さらに表示
Appearance (Color)Dark red to brown
Appearance (Form)Powder
Infrared spectrumConforms
Melting point199°C to 202°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only