6-Chloro-7-iodo-deazapurine, 96%
6-Chloro-7-iodo-deazapurine, 96%
6-Chloro-7-iodo-deazapurine, 96%
Thermo Scientific Chemicals

6-Chloro-7-iodo-deazapurine, 96%

CAS: 123148-78-7 | C6H3ClIN3 | 279.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 448850010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS123148-78-7
IUPAC Name4-chloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Molecular FormulaC6H3ClIN3
InChI KeyCBWBJFJMNBPWAL-UHFFFAOYSA-N
SMILESClC1=C2C(I)=CNC2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point173°C to 180°C
HPLC>=95.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only