TaqMan™ OpenArray™ PGx Express Panel, QuantStudio™ 12K Flex - FAQs
TaqMan™ OpenArray™ PGx Express Panel, QuantStudio™ 12K Flex - FAQs
View additional product information for TaqMan™ OpenArray™ PGx Express Panel, QuantStudio™ 12K Flex - FAQs (4488847)
1 product FAQs found
Do you offer pre-designed TaqMan SNP assay panels?
Pre-designed TaqMan SNP Assay Panels are not available. There are only Pharmacogenomics Drug Metabolism (DME) assays for OpenArray technology.
Find additional tips, troubleshooting help, and resources within our Gene Expression Analysis & Genotyping Support Center.